Merck
  • Chlorine Dioxide is a Better Disinfectant than Sodium Hypochlorite against Multi-Drug Resistant Staphylococcus aureus, Pseudomonas aeruginosa, and Acinetobacter baumannii.
Chlorine Dioxide is a Better Disinfectant than Sodium Hypochlorite against Multi-Drug Resistant Staphylococcus aureus, Pseudomonas aeruginosa, and Acinetobacter baumannii.
Japanese journal of infectious diseases (2015-02-13)
Atsushi Hinenoya, Sharda Prasad Awasthi, Noritomo Yasuda, Ayaka Shima, Hirofumi Morino, Tomoko Koizumi, Toshiaki Fukuda, Takanori Miura, Takashi Shibata, Shinji Yamasaki
ABSTRACT
In this study, we evaluated and compared the antibacterial activity of chlorine dioxide (ClO2) and sodium hypochlorite (NaClO) on various multidrug-resistant strains in the presence of bovine serum albumin and sheep erythrocytes to mimic the blood contamination that frequently occurs in the clinical setting. The 3 most important species that cause nosocomial infections, i.e., methicillin-resistant Staphylococcus aureus (MRSA), multidrug-resistant Pseudomonas aeruginosa (MDRP), and multidrug-resistant Acinetobacter baumannii (MDRA), were evaluated, with three representative strains of each. At a 10-ppm concentration, ClO2 drastically reduced the number of bacteria of all MDRP and MDRA strains, and 2 out of 3 MRSA strains. However, 10 ppm of NaClO did not significantly kill any of the 9 strains tested in 60 seconds (s). In addition, 100 ppm of ClO2 completely killed all MRSA strains, whereas 100 ppm of NaClO failed to significantly lower the number of 2 MRSA strains and 1 MDRA strain. A time-course experiment demonstrated that, within 15 s, 100 ppm of ClO2, but not 100 ppm of NaClO, completely killed all tested strains. Taken together, these data suggest that ClO2 is more effective than NaClO against MRSA, MDRP, and MDRA, and 100 ppm is an effective concentration against these multidrug-resistant strains, which cause fatal nosocomial infections.
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium Thiosulfate Solution, 2 g/dL in deionized water