Merck
  • Taiwanin A incorporated polyurethane fiber sheets for prevention of postoperative cancer recurrence.
Taiwanin A incorporated polyurethane fiber sheets for prevention of postoperative cancer recurrence.
Journal of biomaterials science. Polymer edition (2015-06-03)
Onon Batnyam, Hideyuki Uematsu, Chih-Wei Chou, Shin-Ichiro Suye, Satoshi Fujita
ABSTRACT
In this study, we propose a single solution for prevention of postoperative complications and recurrence of highly metastatic gastrointestinal tract cancers. Here, we demonstrate preparation and characterization of Taiwanin A incorporated polyurethane fiber sheets with excellent mechanical properties and sustained drug release. Sheets with elastic modulus of 8 MPa and ultimate tensile strength of 30 MPa will provide support on surgical staple line preventing leakage at anastomosis. Slight burst release of the drug within 7 days (15%) and further sustained release will inhibit proliferation and migration of remaining cancer cells and maintain locoregional high drug concentration to prevent recurrence of the disease. High elasticity of the material will promote healing process without impeding natural peristalsis movement of gastrointestinal organs.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free