Merck
  • Active immunization against alpha-synuclein ameliorates the degenerative pathology and prevents demyelination in a model of multiple system atrophy.
Active immunization against alpha-synuclein ameliorates the degenerative pathology and prevents demyelination in a model of multiple system atrophy.
Molecular neurodegeneration (2015-04-18)
Markus Mandler, Elvira Valera, Edward Rockenstein, Michael Mante, Harald Weninger, Christina Patrick, Anthony Adame, Sabine Schmidhuber, Radmila Santic, Achim Schneeberger, Walter Schmidt, Frank Mattner, Eliezer Masliah
ABSTRACT
Multiple system atrophy (MSA) is a neurodegenerative disease characterized by parkinsonism, ataxia and dysautonomia. Histopathologically, the hallmark of MSA is the abnormal accumulation of alpha-synuclein (α-syn) within oligodendroglial cells, leading to neuroinflammation, demyelination and neuronal death. Currently, there is no disease-modifying treatment for MSA. In this sense, we have previously shown that next-generation active vaccination technology with short peptides, AFFITOPEs®, was effective in two transgenic models of synucleinopathies at reducing behavioral deficits, α-syn accumulation and inflammation. In this manuscript, we used the most effective AFFITOPE® (AFF 1) for immunizing MBP-α-syn transgenic mice, a model of MSA that expresses α-syn in oligodendrocytes. Vaccination with AFF 1 resulted in the production of specific anti-α-syn antibodies that crossed into the central nervous system and recognized α-syn aggregates within glial cells. Active vaccination with AFF 1 resulted in decreased accumulation of α-syn, reduced demyelination in neocortex, striatum and corpus callosum, and reduced neurodegeneration. Clearance of α-syn involved activation of microglia and reduced spreading of α-syn to astroglial cells. This study further validates the efficacy of vaccination with AFFITOPEs® for ameliorating the neurodegenerative pathology in synucleinopathies.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC