Merck
  • Hedgehog signaling through GLI1 and GLI2 is required for epithelial-mesenchymal transition in human trophoblasts.
Hedgehog signaling through GLI1 and GLI2 is required for epithelial-mesenchymal transition in human trophoblasts.
Biochimica et biophysica acta (2015-04-19)
Chao Tang, Liu Mei, Liyu Pan, Wenyi Xiong, Haibin Zhu, Hongfeng Ruan, Chaochun Zou, Lanfang Tang, Takuma Iguchi, Ximei Wu
ABSTRACT
Epithelial to mesenchymal transition (EMT) is critical for human placental development, trophoblastic differentiation, and pregnancy-associated diseases. Here, we investigated the effects of hedgehog (HH) signaling on EMT in human trophoblasts, and further explored the underlying mechanism. Human primary cytotrophoblasts and trophoblast-like JEG-3 cells were used as in vitro models. Quantitative real-time RT-PCR and Western blot analysis were performed to examine mRNA and protein levels, respectively. Lentiviruses expressing short hairpin RNA were used to knock down the target genes. Reporter assays and chromatin immunoprecipitation were performed to determine the transactivity. Cell migration, invasion and colony formation were accessed by wound healing, Matrigel-coated transwell, and colony formation assays, respectively. Activation of HH signaling induced the transdifferentiation of cytotrophoblasts and trophoblast-like JEG-3 cells from epithelial to mesenchymal phenotypes, exhibiting the decreases in E-Cadherin expression as well as the increases in vimentin expression, invasion, migration and colony formation. Knockdown of GLI1 and GLI2 but not GLI3 attenuated HH-induced transdifferentiation, whereas GLI1 was responsible for the expression of HH-induced key EMT regulators including Snail1, Slug, and Twist, and both GLI1 and GLI2 acted directly as transcriptional repressor of CDH1 gene encoding E-Cadherin. HH through GLI1 and GLI2 acts as critical signals in supporting the physiological function of mature placenta. HH signaling through GLI1 and GLI2 could be required for the maintenance of human pregnancy.
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.