Merck
  • Carbamidomethylation Side Reactions May Lead to Glycan Misassignments in Glycopeptide Analysis.
Carbamidomethylation Side Reactions May Lead to Glycan Misassignments in Glycopeptide Analysis.
Analytical chemistry (2015-05-16)
Zsuzsanna Darula, Katalin F Medzihradszky
ABSTRACT
Iodoacetamide is perhaps the most widely used reagent for the alkylation of free sulfhydryls in proteomic experiments. Here, we report that both incomplete derivatization of Cys side chains and overalkylation of the peptides may lead to the misassignment of glycoforms when LC-MS/MS with electron-transfer dissociation (ETD) alone is used for the structural characterization of glycopeptides. Accurate mass measurements do not help, because the elemental compositions of the misidentified and correct modifications are identical. Incorporation of "higher-energy C-trap dissociation" (HCD), i.e., beam-type collision-induced dissociation data into the database searches with ETD data may prove decisive in most cases. However, the carbamidomethylation of Met residues leads to sulfonium ether formation, and the resulting fixed positive charge triggers a characteristic fragmentation, that eliminates the normal Y1 fragment from the HCD spectra of N-linked glycopeptides, producing an abundant Y1-48 Da ion instead (the nominal mass difference is given relative to the unmodified amino acid sequence), that easily can be mistaken for the side chain loss from Met sulfoxide. In such cases, good quality ETD data may indicate the discrepancy, and will also display abundant fragments due to CH3-S-CH2CONH2 elimination from the charge-reduced precursor ions. Our observations also draw attention to the underreported interference of different unanticipated covalent modifications.
Product Description
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
SAFC
Guanidine hydrochloride
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
SAFC
Iodoacetamide
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
PUGNAc, ≥95% (HPLC)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested