Merck
  • An anion induced multisignaling probe for Hg(2+) and its application for fish kidney and liver tissue imaging studies.
An anion induced multisignaling probe for Hg(2+) and its application for fish kidney and liver tissue imaging studies.
Dalton transactions (Cambridge, England : 2003) (2015-06-26)
Sandip Mandal, Arnab Banerjee, Debasree Ghosh, Dipak Kumar Mandal, Damir A Safin, Maria G Babashkina, Koen Robeyns, Mariusz P Mitoraj, Piotr Kubisiak, Yann Garcia, Debasis Das
ABSTRACT
3',6'-Bis(diethylamino)-2-(pyridin-2-ylmethyl)spiro[isoindoline-1,9'-xanthen]-3-one () was synthesized for the selective fluorescence and colorimetric recognition of Hg(2+) at pH 6.0. In addition, was useful for imaging Hg(2+) in fish kidney and liver tissues using a fluorescence microscope. Spirolactam ring opening of for Hg(2+) recognition is strongly influenced by the nature of the mercury salt and found to be NO3(-)-induced. Other mercury salts such as HgCl2, Hg(CH3COO)2 and Hg(ClO4)2 failed to induce fluorescence and colorimetric response of under the same experimental conditions. For instance, the former salt does not exhibit spirolactam ring opening but forms a new ionic compound (H3L)2[Hg6Cl18]·2H2O (), whose structure has been elucidated by single crystal X-ray diffraction. This might be explained by (1) the higher covalent nature of Hg(2+) and, hence, the lower acidity of the metal center and its inability to induce the ring opening reaction, and (2) the bulky anion, in the case of Hg(ClO4)2, which is also ionic, faces steric hindrance to accommodate within the N(Et)2 group upon spirolactam ring opening.
Sigma-Aldrich
Rhodamine B, Dye content 90 %
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Rhodamine B, ≥95% (HPLC)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Rhodamine B, suitable for fluorescence
Sigma-Aldrich
2-Picolylamine, 99%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%