Merck
  • Polymer brushes interfacing blood as a route toward high performance blood contacting devices.
Polymer brushes interfacing blood as a route toward high performance blood contacting devices.
Macromolecular bioscience (2015-02-04)
František Surman, Tomáš Riedel, Michael Bruns, Nina Yu Kostina, Zdeňka Sedláková, Cesar Rodriguez-Emmenegger
ABSTRACT
In the current study, well-defined polymer brushes are shown as an effective surface modification to resist the adhesion of whole blood and its components. Poly[oligo(ethylene glycol)methylether methacrylate] (poly(MeOEGMA)), poly(hydroxyethyl methacrylate) (poly(HEMA)), poly[N-(2-hydroxypropyl) methacrylamide] (poly(HPMA)), and poly(carboxybetaine acrylamide) (poly(CBAA)) brushes were grown by surface initiated atom transfer radical polymerization (SI-ATRP) and subsequently characterized by Fourier-transform infrared (FTIR) spectroscopy, X-ray photoelectron spectroscopy (XPS), dynamic contact angle measurements, atomic force microscopy (AFM), and surface plasmon resonance (SPR) spectroscopy. All brushes decreased the fouling from blood plasma over 95% and prevented the adhesion of platelets, erythrocytes, and leukocytes as evidenced by SPR and SEM measurements.
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis
Sigma-Aldrich
11-Mercapto-1-undecanol, 99%
Sigma-Aldrich
11-Mercapto-1-undecanol, 97%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
2-Hydroxyethyl methacrylate, ≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
2-Hydroxyethyl methacrylate, contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Sigma-Aldrich
2-Hydroxyethyl methacrylate
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered