Merck
  • Impact of exposure to wood dust on genotoxicity and cytotoxicity in exfoliated buccal and nasal cells.
Impact of exposure to wood dust on genotoxicity and cytotoxicity in exfoliated buccal and nasal cells.
Mutagenesis (2015-05-11)
Georg Wultsch, Armen Nersesyan, Michael Kundi, Karl-Heinz Wagner, Franziska Ferk, Robert Jakse, Siegfried Knasmueller
ABSTRACT
Wood dust was classified by the IARC as a human carcinogen which causes sinonasal tumours. However, the exposure in different industries varies strongly and the risks of workers depend on the specific situation which can be assessed by the use of biomonitoring methods. The aim of this study was to investigate the workers who are exposed to low dust levels (below the permitted concentrations) with cytogenetic and biochemical methods. Micronuclei (MNi) which are indicative for genomic damage, nuclear buds which reflect gene amplification, binucleated cells which are caused by mitotic disturbances and acute cytotoxicity parameters (pyknosis, karyorrhexis, condensed chromatin, karyolysis) were monitored in buccal and nasal cells of workers of a veneer factory (n = 51) who are exposed to volatile wood-derived compounds, in carpenters of a furniture factory which use no synthetic chemicals (n=38) and in a control group (n = 65). Additionally, markers were measured in blood plasma which reflect inflammations (C-reactive protein, CRP) and the redox status, namely malondialdehyde (MDA) and oxidised low density proteins (oxLDL). No induction of micronucleated cells was observed in both epithelia in the two exposure groups while all other nuclear anomalies except pyknosis were increased; also one health-related biochemical marker (MDA) was significantly elevated in the workers. Taken together, the results of our study show that exposure to low levels of wood dust does not cause formation of MNi indicating that the cancer risks of the workers are not increased as a consequence of genetic damage while positive results were obtained in earlier studies with workers who are exposed to high dust levels. However, our findings indicate that wood dust causes cytotoxic effects which may lead to inflammations.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
4,4′-Diaminodiphenylmethane, ≥97.0% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether