Merck
  • Taibaiella yonginensis sp. nov., a bacterium isolated from soil of Yongin city.
Taibaiella yonginensis sp. nov., a bacterium isolated from soil of Yongin city.
Antonie van Leeuwenhoek (2015-06-14)
Hina Singh, Juan Du, KyungHwa Won, Jung-Eun Yang, Shahina Akter, Ki-Young Kim, ChangShik Yin, Tae-Hoo Yi
ABSTRACT
A novel bacterial strain, designated as THG-SC4(T), was isolated from a soil sample collected from Yongin city in South Korea. Cells of the strain were Gram-negative, aerobic, rod-shaped and non-motile. The strain grew optimally at 28-30 °C; at pH 7.0 and in the absence of NaCl. Flexirubin-type pigments were found to be present. On the basis of 16S rRNA gene sequence similarity, strain THG-SC4(T) was shown to belong to the genus Taibaiella and shares high sequence similarity with Taibaiella smilacinae KCTC 32316(T) (95.4 %), followed by Taibaiella koreensis KACC 17171(T) (94.3 %) and Taibaiella chishuiensis JCM 19637(T) (94.2 %). The DNA G+C content of the novel isolate was determined to be 43.1 mol% and MK-7 was identified as the predominant isoprenoid quinone. The only polyamine was homospermidine. The major polar lipids were phosphatidylethanolamine, an unidentified aminophospholipid and an unidentified polar lipid. The predominant fatty acids were identified as iso-C15:0, iso-C15:1 G, C16:0 and iso-C17:03-OH. On the basis of data from this polyphasic taxonomic study, strain THG-SC4(T) is considered to represent a novel species of the genus Taibaiella, for which the name Taibaiella yonginensis sp. nov. is proposed. The type strain is THG-SC4(T) (=KACC 18372(T) = CCTCC AB 2014316(T)).
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Supelco
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
SAFC
L-Tyrosine
Sigma-Aldrich
Ninhydrin, ACS reagent
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Phosphomolybdic Acid Solution
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from Escherichia coli strain B, Genomic, unsheared
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis