Merck
  • Enteric glial cells are associated with stress-induced colonic hyper-contraction in maternally separated rats.
Enteric glial cells are associated with stress-induced colonic hyper-contraction in maternally separated rats.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2015-05-12)
Y Fujikawa, K Tominaga, F Tanaka, T Tanigawa, T Watanabe, Y Fujiwara, T Arakawa
ABSTRACT
Enteric glial cells (EGCs) play important roles in enteric integrity and regulation of gastrointestinal function. However, whether EGCs undergo pathophysiological changes in stress-associated gastrointestinal disorders is unknown. We investigated structural and functional alterations in colonic EGCs and their roles in colonic contraction in an irritable bowel syndrome (IBS) model. As a chronic stress, male Wistar rats underwent 3-h maternal separation during postnatal days 2-14. As an acute stress, we used water-immersion stress (4 h) in adulthood (at 8 weeks). We quantitatively and morphologically evaluated enteric neurons and EGCs using whole-mount longitudinal muscle-myenteric plexus preparations. Colonic contraction was analyzed with electrical field stimulation (EFS). Glial fibrillary acidic protein (GFAP) expression and the number of total, cholinergic, and nitrergic neurons were unchanged in maternally separated rats with acute stress (combined stress: an IBS model) compared with controls. However, the density of GFAP-positive EGC processes that apparently overlapped with the neurons and the extent of bulbous swelling of terminals increased according to the stress intensity: control, acute stress, maternal separation, and combined stress. EFS-induced colonic contractions were significantly greater in the combined stress rats than in controls. Higher dose of fluorocitrate, a selective inhibitor of EGC metabolism, was required to inhibit both EFS-induced contraction and EGCs activation in the combined stress rats than in controls. Colonic EGCs exhibited structural alterations according to the stress intensity. EGCs were associated with stress-induced colonic hyper-contraction in the combined stress rats, which may underlie the pathogenesis of IBS.
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)