Merck
  • Simple and sensitive progesterone detection in human serum using a CdSe/ZnS quantum dot-based direct binding assay.
Simple and sensitive progesterone detection in human serum using a CdSe/ZnS quantum dot-based direct binding assay.
Analytical biochemistry (2015-05-13)
Sung-Duk Oh, Hong Dinh Duong, Jong Il Rhee
ABSTRACT
In this study, we developed a CdSe/ZnS quantum dot (QD)-based immunoassay for use in determining the presence of progesterone (P4) in human serum. Hydrophilic QDs were conjugated to anti-progesterone antibody (P4Ab) via ethyl-3-(dimethylaminopropyl)carbodiimide (EDC) and N-hydroxysuccinimide (NHS) as coupling reagents. After purification, the P4Ab-QD conjugates were immobilized onto the wells of a 96-well microtiter plate, and a direct-binding immunoassay based on the binding of P4 to immobilized P4Ab-QD conjugates had a detection limit of 0.21 ng/ml and a sensitivity of 1.37 ng/ml, with a linear range of 0.385 to 4.55 ng/ml. The proposed immunoassay was successfully used to determine the P4 concentration in real human serum, and the results showed a good correlation with the accredited radioimmunoassay (RIA).
Product Description
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Progesterone, ≥99%
Cadmium(II) acetate, SAFC Hitech®, anhydrous, 99.995%
Sigma-Aldrich
Cadmium(II) acetate, anhydrous, 99.995%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Trioctylphosphine, technical grade, 90%
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Hexadecylamine, 98%
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
(3-Glycidyloxypropyl)trimethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
Sigma-Aldrich
2-Mercaptopropionic acid, ≥95%, FG
Sigma-Aldrich
N-Hydroxysuccinimide, 98%