Merck
  • Oxidative modification induced by photodynamic therapy with Photofrin®II and 2-methoxyestradiol in human ovarian clear carcinoma (OvBH-1) and human breast adenocarcinoma (MCF-7) cells.
Oxidative modification induced by photodynamic therapy with Photofrin®II and 2-methoxyestradiol in human ovarian clear carcinoma (OvBH-1) and human breast adenocarcinoma (MCF-7) cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2015-05-12)
Jolanta Saczko, Anna Choromańska, Nina Rembiałkowska, Magda Dubińska-Magiera, Iwona Bednarz-Misa, Julia Bar, Anna Marcinkowska, Julita Kulbacka
ABSTRACT
Ovarian cancer is among the most lethal cancers in women. The successful anticancer treatment depends on the effectiveness of cytotoxic effect of applied therapeutic procedures either alone or in combination with other treatments. Photodynamic therapy (PDT) is a relatively new method of anticancer therapy. Its dominant mechanism of action is the over-production of reactive oxygen species induced by oxidative stress in malignant cells, which attack lipid membranes, proteins and nucleic acids. One of the important mechanisms is induction of unfolded protein response, ubiquitin-proteasome pathway of protein degradation. The aim of this study was to evaluate the cytotoxic effect of various protective enzymes in ovarian carcinoma clear cell line in comparison to the model breast cell line after photodynamic reaction and photodynamic reaction with 2-methoxyestradiol (2-Me). Human malignant ovarian cell line (OvBH-1) was used and human breast adenocarcinoma cells (MCF-7) were used as a control. Photodynamic reaction (PDR) with Photofrin(®)II and Ph(®)II with 2-Me was performed. The expression of protective proteins by immunocytochemistry (HSP70 and iNOS) and western blot (Hsp27 and Hsp70) methods was evaluated directly, 3 and 6 h after PDR. The changes in cells' cytoskeleton were evaluated using immunofluorescence by confocal microscopy. The expression of iNOS was observed for both experiments with differential intensity and quantity. A higher expression of Hsp70 in MCF-7 cells was observed than in OvBh-1 cells. The reorganization of cytoskeleton and nucleus was observed after 3 and 6 h after exposition to light.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Glycine
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)