Merck
  • Topography-Guided Proliferation: Distinct Surface Microtopography Increases Proliferation of Chondrocytes In Vitro.
Topography-Guided Proliferation: Distinct Surface Microtopography Increases Proliferation of Chondrocytes In Vitro.
Tissue engineering. Part A (2015-09-29)
Natasja Leth Joergensen, Dang Quang Svend Le, Ole Zoffmann Andersen, Morten Foss, Carl Christian Danielsen, Casper Bindzus Foldager, Martin Lind, Helle Lysdahl
ABSTRACT
Chondrocyte-based cartilage repair techniques require control of articular chondrocyte expansion ex vivo. Articular chondrocytes have limited availability, and prolonged culturing to obtain a cell number sufficient for clinical use often results in phenotypic alterations and increased costs. In this study, we applied a screening library consisting of micrometer-sized topographical features, termed biosurface structure array (BSSA), to identify specific topographical microstructures affecting the proliferation of human chondrocytes in passage 1 (P1) or 2 (P2). The BSSA library comprised 10 patterns and 16 combinations of pillar size (X) and interpillar gap size (Y). Specific microstructures significantly increased the chondrocytes' proliferative responsiveness in term of patterns, X and Y for P2 compared with P1. The P1 and P2 chondrocytes responded independently to similar patterns after 4 days of culturing, whereas only chondrocytes at P2 responded to specific microstructures with Y = 1 μm and X = 2, 4 μm by a 2.3- and 4.4-fold increased proliferation, respectively. In conclusion, these findings indicate that specific surface topographies promote chondrocyte proliferation and may, indeed, be a tool to control the behavior of chondrocytes in vitro.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
1H,1H,2H,2H-Perfluorooctyltriethoxysilane, 98%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)