Merck
  • Glycosylation-dependent interaction between CD69 and S100A8/S100A9 complex is required for regulatory T-cell differentiation.
Glycosylation-dependent interaction between CD69 and S100A8/S100A9 complex is required for regulatory T-cell differentiation.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-08-25)
Chih-Ru Lin, Tong-You Wade Wei, Hsien-Yu Tsai, Ying-Ta Wu, Pei-Yu Wu, Shui-Tein Chen
ABSTRACT
Cluster of differentiation (CD)69 is a leukocyte activation receptor involved in the maintenance of immune homeostasis and is positively selected in activated regulatory T (Treg) cells, implicating its role during Treg-cell differentiation. By RNA interference, we show that CD69 is not sufficient to support the conversion of CD4(+) naive T cells into Treg cells, whereas it does that of human peripheral blood mononuclear cells (hPBMCs) (P < 0.01), suggesting that a ligand-receptor interaction is required for CD69 function. Using immunoprecipitation and mass spectrometry, we identified the S100A8/S100A9 complex as the natural ligand of CD69 in hPBMCs. CD69 specifically associates with S100A8/S100A9 complex as confirmed by in vitro binding and competition assay, and the treatment of CD69 with peptide-N-glycosidase significantly abolishes such association. In agreement, the glycomics analysis determines the glycosylation site and the N-glycan composition of CD69, and terminal removal of sialic acid from that N-linked glycans reverses the generation of forkhead box P3-positive Treg cells (23.21%; P < 0.05). More specifically, we showed that CD69-S100A8/S100A9 association is required for the up-regulation of suppressor of cytokine signaling 3 resulting in inhibited signaling of signal transducer and activator of transcription 3 (36.54% increase upon CD69 silencing; P < 0.01). This might in turn support the secretion of key regulator TGF-β (∼ 3.28-fold decrease upon CD69 silencing; P < 0.05), leading to reduced production of IL-4 in hPBMCs. Our results demonstrate the functional and mechanistic interplays between CD69 and S100A8/S100A9 in supporting Treg-cell differentiation.
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
PIPES, BioXtra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%