Merck
  • Ultra trace analysis of PAHs by designing simple injection of large amounts of analytes through the sample reconcentration on SPME fiber after magnetic solid phase extraction.
Ultra trace analysis of PAHs by designing simple injection of large amounts of analytes through the sample reconcentration on SPME fiber after magnetic solid phase extraction.
Talanta (2015-11-26)
Nader Khodaee, Ali Mehdinia, Reyhaneh Esfandiarnejad, Ali Jabbari
ABSTRACT
A simple solventless injection method was introduced based on the using of a solid-phase microextraction (SPME) fiber for injection of large amounts of the analytes extracted by the magnetic solid phase extraction (MSPE) procedure. The resulted extract from MSPE procedure was loaded on a G-coated SPME fiber, and then the fiber was injected into the gas chromatography (GC) injection port. This method combines the advantages of exhaustive extraction property of MSPE and the solvent-less injection of SPME to improve the sensitivity of the analysis. In addition, the analytes were re-concentrated prior to inject into the gas chromatography (GC) inlet because of the organic solvent removing from the remaining extract of MSPE technique. Injection of the large amounts of analytes was made possible by using the introduced procedure. Fourteen polycyclic aromatic hydrocarbons (PAHs) with different volatility were used as model compounds to investigate the method performance for volatile and semi-volatile compounds. The introduced method resulted in the higher enhancement factors (5097-59376), lower detection limits (0.29-3.3pgmL(-1)), and higher sensitivity for the semi-volatile compounds compared with the conventional direct injection method.
Sigma-Aldrich
Thiophene, ≥99%
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, puriss., ≥99%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3