Merck
  • Annexin A2 regulates angiogenesis and invasion phenotypes of malignant glioma.
Annexin A2 regulates angiogenesis and invasion phenotypes of malignant glioma.
Brain tumor pathology (2015-02-24)
Manabu Onishi, Tomotsugu Ichikawa, Kazuhiko Kurozumi, Satoshi Inoue, Tomoko Maruo, Yoshihiro Otani, Kentaro Fujii, Joji Ishida, Yosuke Shimazu, Koichi Yoshida, Hiroyuki Michiue, E Antonio Chiocca, Isao Date
ABSTRACT
We have established a pair of animal models (J3T-1 and J3T-2) with different invasive and angiogenic phenotypes, and demonstrated that annexin A2 is expressed at higher levels in J3T-1 than J3T-2 cells. The function of annexin A2 in relation to angiogenesis and invasion was investigated using these models. Stable silencing or overexpression of annexin A2 in J3T-1 and J3T-2 cells (J3T-1shA and J3T-2A cells) was established and used. Thirty human glioblastoma samples were evaluated for expression of annexin A2, vascular endothelial growth factor (VEGF) and platelet-derived growth factor (PDGF). Immunohistochemical and quantitative reverse-transcription polymerase chain reaction analyses revealed higher expression of annexin A2, VEGF and PDGF in J3T-1 and J3T-2A cells. Cultured J3T-1 and J3T-2A cells exhibited higher adhesive ability to endothelial cells. Histopathological analysis of animal brain tumors revealed that J3T-1 and J3T-2A tumors displayed marked angiogenesis and invasion along the neovasculature, whereas J3T-2 and J3T-1shA tumors exhibited diffuse, infiltrative invasion without angiogenesis. Positive expression of annexin A2 was observed in tumor cells surrounding dilated vessels in 25/30 human glioblastoma specimens. Our results reveal that the phenotype of glioma invasion is closely related to angiogenesis. We identify annexin A2 as a factor regulating angiogenesis and invasion of malignant gliomas.
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Anxa2
Sigma-Aldrich
MISSION® esiRNA, targeting human ANXA2