Merck
  • Phenolic compounds content and antioxidant capacity of meals provided to elementary public schools in Chile during 2011.
Phenolic compounds content and antioxidant capacity of meals provided to elementary public schools in Chile during 2011.
Archivos latinoamericanos de nutricion (2016-01-29)
Mariane Lutz, Romina Barraza, Carolina Henríquez
ABSTRACT
The Chilean National School Feeding Program (NSFP) delivers breakfast and lunch meals that supply 250 and 450 kcal, respectively, along the country. In the last decades, a significant increase of obesity has been observed in primary education children, and it involves risk factors of non-communicable diseases. The dietary intake of foods containing phenolic compounds (PC) exerts favorable effects on health by reducing risk factors of prevalent diseases. The aim of the study was to measure the PC content and antioxidant capacity (AC) [ORAC and DPPH] of meals provided by the NSFP in Quillota, Chile, in 2011. The PC supply of the whole meals served ranged from 362.7 to 1,730 mg GAE. The best breakfast foods include whole grain cookie (2.59 ± 0.3 mg GAE/g), bread with avocado, quince jelly or strawberry jam (1.61 ± 0.13 to 2.05 ± 0.3 mg GAE/g); while the best lunch salads include beetroot, lettuce, and cabbage/fish (1.66 ± 0.3 to 2.35 ± 0.1 mg GAE/g), and main courses contain legumes, or mixed vegetables. The lowest PC contents were observed in pasta and rice preparations (p < 0.05). Among desserts, the best source of PC is fruit (1.81 ± 0.04 to 6.91 ± 0.31 mg GAE/g). The correlation between PC and AC varied according to the type of meal. PC content and AC are additional criteria for selecting the best quality meals, in addition to the nutrients and energy content. The results support the recommendation to increase the supply of fruits and vegetable/legumes preparations and fruits instead of starchy foods to scholars.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%