Merck
  • Differential susceptibility of brain regions to tributyltin chloride toxicity.
Differential susceptibility of brain regions to tributyltin chloride toxicity.
Environmental toxicology (2014-06-05)
Sumonto Mitra, Waseem A Siddiqui, Shashi Khandelwal
ABSTRACT
Tributyltin (TBT), a well-known endocrine disruptor, is an omnipresent environmental pollutant and is explicitly used in many industrial applications. Previously we have shown its neurotoxic potential on cerebral cortex of male Wistar rats. As the effect of TBT on other brain regions is not known, we planned this study to evaluate its effect on four brain regions (cerebellum, hippocampus, hypothalamus, and striatum). Four-week-old male Wistar rats were gavaged with a single dose of TBT-chloride (TBTC) (10, 20, and 30 mg/kg) and sacrificed on days 3 and 7, respectively. Effect of TBTC on blood-brain barrier (BBB) permeability and tin (Sn) accumulation were measured. Oxidative stress indexes such as reactive oxygen species (ROS), reduced and oxidized glutathione (GSH/GSSG) ratio, lipid peroxidation, and protein carbonylation were analyzed as they play an imperative role in various neuropathological conditions. Since metal catalyzed reactions are a major source of oxidant generation, levels of essential metals like iron (Fe), zinc (Zn), and calcium (Ca) were estimated. We found that TBTC disrupted BBB and increased Sn accumulation, both of which appear significantly correlated. Altered metal homeostasis and ROS generation accompanied by elevated lipid peroxidation and protein carbonylation indicated oxidative damage which appeared more pronounced in the striatum than in cerebellum, hippocampus, and hypothalamus. This could be associated to the depleted GSH levels in striatum. These results suggest that striatum is more susceptible to TBTC induced oxidative damage as compared with other brain regions under study.
Sigma-Aldrich
Guanidine hydrochloride, ≥99% (titration), organic base and chaeotropic agent
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
2,4-Dinitrophenylhydrazine, reagent grade, 97%
Supelco
Formamide solution, suitable for NMR (reference standard), 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Tributyltin chloride, 96%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%