Merck
  • How relevant are assembled equilibrium samples in understanding structure formation during lipid digestion?
How relevant are assembled equilibrium samples in understanding structure formation during lipid digestion?
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-07-28)
Stephanie Phan, Stefan Salentinig, Adrian Hawley, Ben J Boyd
ABSTRACT
Lipid-based formulations are gaining interest for use as drug delivery systems for poorly water-soluble drug compounds. During digestion, the lipolysis products self-assemble with endogenous surfactants in the gastrointestinal tract to form colloidal structures, enabling enhanced drug solubilisation. Although earlier studies in the literature focus on assembled equilibrium systems, little is known about structure formation under dynamic lipolysis conditions. The purpose of this study was to investigate the likely colloidal structure formation in the small intestine after the ingestion of lipids, under equilibrium and dynamic conditions. The structural aspects were studied using small angle X-ray scattering and dynamic light scattering, and were found to depend on lipid composition, lipid chain length, prandial state and emulsification. Incorporation of phospholipids and lipolysis products into bile salt micelles resulted in swelling of the structure. At insufficient bile salt concentrations, a co-existing lamellar phase was observed, due to a reduction in the solubilisation capacity for lipolysis products. Emulsification accelerated the rate of lipolysis and structure formation.
Sigma-Aldrich
Dodecanoic acid, ≥99% (GC/titration)
Sigma-Aldrich
Dodecanoic acid, 98%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
1-Oleoyl-rac-glycerol, technical, ~40% (TLC)
Sigma-Aldrich
1-Octanoyl-rac-glycerol, ≥99%
Sigma-Aldrich
1-Oleoyl-rac-glycerol, ≥99%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Octanoic acid, ≥99%