Merck
  • Comparison of twin-cell centrifugal partition chromatographic columns with different cell volume.
Comparison of twin-cell centrifugal partition chromatographic columns with different cell volume.
Journal of chromatography. A (2015-06-25)
Johannes Goll, Gregoire Audo, Mirjana Minceva
ABSTRACT
Two twin-cell centrifugal partition chromatographic columns (SCPC 250 and SCPE-250-BIO, Armen Instrument, France) with the same column volume but different cell size and number were compared in terms of stationary phase retention and column efficiency. The columns were tested with two types of solvent systems: a commonly used organic solvent based biphasic system from the ARIZONA solvent system family and a polymer/salt based aqueous two phase system (ATPS). The efficiency of the columns was evaluated by pulse injection experiments of two benzenediols (pyrocatechol and hydroquinone) in the case of the ARIZONA system and a protein mixture (myoglobin and lysozyme) in the case of the ATPS. As result of high stationary phase retention, the column with the lower number of larger twin-cells (SCPE-250-BIO) is suitable for protein separations using ATPS. On the other hand, due to higher column efficiency, the column with the greater number of smaller cells (SCPC 250) is superior for batch elution separations performed with standard liquid-liquid chromatography organic solvent based biphasic systems.
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Hydroquinone, meets USP testing specifications
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, 99%
Sigma-Aldrich
Hydroquinone, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl acetate
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)