Merck
  • Transient inhibition of interleukin 4 signaling by T cell receptor ligation.
Transient inhibition of interleukin 4 signaling by T cell receptor ligation.
The Journal of experimental medicine (2000-10-18)
J Zhu, H Huang, L Guo, T Stonehouse, C J Watson, J Hu-Li, W E Paul
ABSTRACT
Interleukin (IL)-4 and IL-12 together with T cell receptor (TCR) engagement are crucial for the differentiation of CD4(+) T cells into T helper (Th)2 or Th1 cells, respectively. Although IL-4 receptors (IL-4Rs) but not IL-12Rs are expressed on naive CD4(+) T cells, IL-4 has no apparent advantage over IL-12 in driving naive T cell differentiation when the cells are primed with both IL-4 and IL-12 in vitro. It was found that IL-4-induced phosphorylation of Janus kinases 1 and 3, IL-4R alpha, signal transducer and activator of transcription 6, and insulin receptor substrate 2 was strikingly but transiently inhibited by TCR ligation both in conventional and TCR transgenic T cells. TCR engagement also blocked the expression of an IL-4-inducible gene. Signals induced by other cytokines, including IL-2, IL-6, and interferon alpha, but not by insulin-like growth factor 1, were also blocked by TCR engagement. The capacity of various inhibitors to reverse TCR-mediated inhibition of IL-4 signaling suggested that activation of the Ras-mitogen-activated protein kinase pathway and of the calcineurin pathway contribute to desensitizing IL-4R. IL-4 responsiveness returned at about the time ( approximately 12 h) that IL-12-mediated signaling was first observed. Thus, through different mechanisms, neither IL-4R nor IL-12R has any clear advantage in polarizing cells; rather, the availability of cytokine is probably the limiting factor in this process.
Sigma-Aldrich
Anti-JAK1 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-JAK3 Antibody, clone B32-32, clone B32-32, Upstate®, from mouse
Sigma-Aldrich
Anti-JAK3 Antibody, serum, Upstate®