Merck
  • Inhibition of histone deacetylase 3 causes replication stress in cutaneous T cell lymphoma.
Inhibition of histone deacetylase 3 causes replication stress in cutaneous T cell lymphoma.
PloS one (2013-07-31)
Christina E Wells, Srividya Bhaskara, Kristy R Stengel, Yue Zhao, Bianca Sirbu, Benjamin Chagot, David Cortez, Dineo Khabele, Walter J Chazin, Andrew Cooper, Vincent Jacques, James Rusche, Christine M Eischen, Laura Y McGirt, Scott W Hiebert
ABSTRACT
Given the fundamental roles of histone deacetylases (HDACs) in the regulation of DNA repair, replication, transcription and chromatin structure, it is fitting that therapies targeting HDAC activities are now being explored as anti-cancer agents. In fact, two histone deacetylase inhibitors (HDIs), SAHA and Depsipeptide, are FDA approved for single-agent treatment of refractory cutaneous T cell lymphoma (CTCL). An important target of these HDIs, histone deacetylase 3 (HDAC3), regulates processes such as DNA repair, metabolism, and tumorigenesis through the regulation of chromatin structure and gene expression. Here we show that HDAC3 inhibition using a first in class selective inhibitor, RGFP966, resulted in decreased cell growth in CTCL cell lines due to increased apoptosis that was associated with DNA damage and impaired S phase progression. Through isolation of proteins on nascent DNA (iPOND), we found that HDAC3 was associated with chromatin and is present at and around DNA replication forks. DNA fiber labeling analysis showed that inhibition of HDAC3 resulted in a significant reduction in DNA replication fork velocity within the first hour of drug treatment. These results suggest that selective inhibition of HDAC3 could be useful in treatment of CTCL by disrupting DNA replication of the rapidly cycling tumor cells, ultimately leading to cell death.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Bexarotene, ≥98% (HPLC)
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse