Merck
  • Cationic palladium(II)-catalyzed addition of arylboronic acids to nitriles. One-step synthesis of benzofurans from phenoxyacetonitriles.
Cationic palladium(II)-catalyzed addition of arylboronic acids to nitriles. One-step synthesis of benzofurans from phenoxyacetonitriles.
Organic letters (2006-12-15)
Baowei Zhao, Xiyan Lu
ABSTRACT
[Structure: see text] A cationic palladium complex catalyzed addition of arylboronic acids to nitriles to yield aryl ketones with moderate to good yields was developed. A one-step synthesis of benzofurans from phenoxyacetonitriles under the catalysis of [(bpy)Pd+(micro-OH)]2(-OTf)2 or [(bpy)Pd2+(H2O)2](-OTf)2 was developed which showed that the cationic palladium catalyst is highly active for these addition reactions.
Sigma-Aldrich
2-Naphthylboronic acid, ≥95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Fluorophenylboronic acid, ≥95%
Pricing and availability is not currently available.