Merck
  • Suppressor of IKKɛ is an essential negative regulator of pathological cardiac hypertrophy.
Suppressor of IKKɛ is an essential negative regulator of pathological cardiac hypertrophy.
Nature communications (2016-06-02)
Ke-Qiong Deng, Aibing Wang, Yan-Xiao Ji, Xiao-Jing Zhang, Jing Fang, Yan Zhang, Peng Zhang, Xi Jiang, Lu Gao, Xue-Yong Zhu, Yichao Zhao, Lingchen Gao, Qinglin Yang, Xue-Hai Zhu, Xiang Wei, Jun Pu, Hongliang Li
ABSTRACT
Although pathological cardiac hypertrophy represents a leading cause of morbidity and mortality worldwide, our understanding of the molecular mechanisms underlying this disease is still poor. Here, we demonstrate that suppressor of IKKɛ (SIKE), a negative regulator of the interferon pathway, attenuates pathological cardiac hypertrophy in rodents and non-human primates in a TANK-binding kinase 1 (TBK1)/AKT-dependent manner. Sike-deficient mice develop cardiac hypertrophy and heart failure, whereas Sike-overexpressing transgenic (Sike-TG) mice are protected from hypertrophic stimuli. Mechanistically, SIKE directly interacts with TBK1 to inhibit the TBK1-AKT signalling pathway, thereby achieving its anti-hypertrophic action. The suppression of cardiac remodelling by SIKE is further validated in rats and monkeys. Collectively, these findings identify SIKE as a negative regulator of cardiac remodelling in multiple animal species due to its inhibitory regulation of the TBK1/AKT axis, suggesting that SIKE may represent a therapeutic target for the treatment of cardiac hypertrophy and heart failure.
Sigma-Aldrich
Anti-α-Actinin Antibody, clone AT6/172, clone AT6/172, Upstate®, from mouse
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks