Merck
  • Ink4a/Arf and oncogene-induced senescence prevent tumor progression during alternative colorectal tumorigenesis.
Ink4a/Arf and oncogene-induced senescence prevent tumor progression during alternative colorectal tumorigenesis.
Cancer cell (2010-08-17)
Moritz Bennecke, Lydia Kriegl, Monther Bajbouj, Kristin Retzlaff, Sylvie Robine, Andreas Jung, Melek C Arkan, Thomas Kirchner, Florian R Greten
ABSTRACT
Colonic cancers with a serrated morphology have been proposed to comprise a molecularly distinct tumor entity following an alternative pathway of genetic alterations independently of APC mutations. We demonstrate that intestinal epithelial cell specific expression of oncogenic K-ras(G12D) in mice induces serrated hyperplasia, which is characterized by p16(ink4a) overexpression and induction of senescence. Deletion of Ink4a/Arf in K-ras(G12D) expressing mice prevents senescence and leads to invasive, metastasizing carcinomas with morphological and molecular alterations comparable to human KRAS mutated serrated tumors. Thus, we suggest that oncogenic K-ras represents a key player during an alternative, serrated pathway to colorectal cancer and hence propose RAS-RAF-MEK signaling apart from APC as an additional gatekeeper in colorectal tumor development.
Sigma-Aldrich
Ras Activation Assay Kit
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid
Sigma-Aldrich
Ral Activation Assay Kit, Non-radioactive Ral Activation Assay Kit can be used to precipitate Ral-A-GTP from cell lysates & detection by a Ral-A specific antibody.