Merck
  • Characterization of osimertinib (AZD9291)-resistant non-small cell lung cancer NCI-H1975/OSIR cell line.
Characterization of osimertinib (AZD9291)-resistant non-small cell lung cancer NCI-H1975/OSIR cell line.
Oncotarget (2016-11-12)
Zheng-Hai Tang, Xiao-Ming Jiang, Xia Guo, Chi Man Vivienne Fong, Xiuping Chen, Jin-Jian Lu
ABSTRACT
Osimertinib (OSI, also known as AZD9291) is the newest FDA-approved epidermal growth factor receptor (EGFR) tyrosine kinase inhibitor for non-small cell lung cancer (NSCLC) patients with EGFR T790M mutation. However, resistance to OSI is likely to progress and the study of potential OSI-resistant mechanisms in advanced is necessary. Here, the OSI-resistant NCI-H1975/OSIR cells were established. After cells developed resistance to OSI, cell proliferation was decreased while cell migration and invasion were increased. The NCI-H1975/OSIR cells exhibited more resistance to gefitinib, erlotinib, afatinib, rociletinib, doxorubicin, and fluorouracil, meanwhile showing higher sensitivity to paclitaxel, when compared with NCI-H1975 cells. In addition, the NCI-H1975/OSIR cells did not display multidrug resistance phenotype. The activation and expression of EGFR were decreased after cells exhibited resistance. Compared with NCI-H1975 cells, the activation of ERK and AKT in NCI-H1975/OSIR cells could not be significantly inhibited by OSI treatment. Navitoclax (ABT-263)-induced cell viability inhibition and apoptosis were more significant in NCI-H1975/OSIR cells than that in NCI-H1975 cells. Moreover, these effects of navitoclax in NCI-H1975/OSIR cells could be reversed by pretreatment of Z-VAD-FMK. Collectively, loss of EGFR could pose as one of the OSI-resistant mechanisms and navitoclax might be the candidate drug for OSI-resistant NSCLC patients.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder