Merck
Foldable and Disposable Memory on Paper.
Scientific reports (2016-12-07)
Byung-Hyun Lee, Dong-Il Lee, Hagyoul Bae, Hyejeong Seong, Seung-Bae Jeon, Myung-Lok Seol, Jin-Woo Han, M Meyyappan, Sung-Gap Im, Yang-Kyu Choi
ABSTRACT
Foldable organic memory on cellulose nanofibril paper with bendable and rollable characteristics is demonstrated by employing initiated chemical vapor deposition (iCVD) for polymerization of the resistive switching layer and inkjet printing of the electrode, where iCVD based on all-dry and room temperature process is very suitable for paper electronics. This memory exhibits a low operation voltage of 1.5 V enabling battery operation compared to previous reports and wide memory window. The memory performance is maintained after folding tests, showing high endurance. Furthermore, the quick and complete disposable nature demonstrated here is attractive for security applications. This work provides an effective platform for green, foldable and disposable electronics based on low cost and versatile materials.
Sigma-Aldrich
Luperox® DI, tert-Butyl peroxide, 98%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor