Merck
  • Beta-asarone protects against MPTP-induced Parkinson's disease via regulating long non-coding RNA MALAT1 and inhibiting α-synuclein protein expression.
Beta-asarone protects against MPTP-induced Parkinson's disease via regulating long non-coding RNA MALAT1 and inhibiting α-synuclein protein expression.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-10-25)
Qi-Shun Zhang, Zhao-Hui Wang, Jian-Lei Zhang, Yan-Li Duan, Guo-Fei Li, Dong-Lin Zheng
ABSTRACT
Numerous long non-coding RNAs (lncRNA) have been identified in neurodegenerative disorders including Parkinson's disease (PD). Emerging evidence demonstrates that β-asarone functions as neuroprotective effects in both in vitro and in vivo models. However, the role of β-asarone and its potential mechanism in PD remain not completely clear. MPTP-induced PD mouse model and SH-SY5Y cells subjected to MPP+ as its in vitro model were used to evaluate the effects of β-asarone on PD. LncRNA MALAT1 and α-synuclein expression were determined by real-time PCR and western blot methods. β-Asarone significantly increased the TH+ cells number and decreased the expression levels of MALAT1 and α-synuclein in midbrain tissue of PD mice. RNA pull-down and immunoprecipitation assays confirmed that MALAT1 associated with α-synuclein, leading to the increased stability of α-synuclein and its expression in SH-SY5Y cells. β-asarone elevated the viability of cells exposed to MPP+. Either overexpressed MALAT1 or α-synuclein could canceled the protective effect of β-asarone on cell viability. In PD mice, pcDNA-MALAT1 also decreased the TH+ cells number and increased the α-synuclein expression in PD mice with treatment of β-asarone. β-Asarone functions as a neuroprotective effect in both in vivo and in vitro models of PD via regulating MALAT1 and α-synuclein expression.