Merck
  • The anti-inflammatory modulatory role of Solidago chilensis Meyen in the murine model of the air pouch.
The anti-inflammatory modulatory role of Solidago chilensis Meyen in the murine model of the air pouch.
The Journal of Pharmacy and Pharmacology (2008)
Liz R