Merck
  • Lysyl oxidase drives tumour progression by trapping EGF receptors at the cell surface.
Lysyl oxidase drives tumour progression by trapping EGF receptors at the cell surface.
Nature communications (2017-04-19)
HaoRan Tang, Leo Leung, Grazia Saturno, Amaya Viros, Duncan Smith, Gianpiero Di Leva, Eamonn Morrison, Dan Niculescu-Duvaz, Filipa Lopes, Louise Johnson, Nathalie Dhomen, Caroline Springer, Richard Marais
ABSTRACT
Lysyl oxidase (LOX) remodels the tumour microenvironment by cross-linking the extracellular matrix. LOX overexpression is associated with poor cancer outcomes. Here, we find that LOX regulates the epidermal growth factor receptor (EGFR) to drive tumour progression. We show that LOX regulates EGFR by suppressing TGFβ1 signalling through the secreted protease HTRA1. This increases the expression of Matrilin2 (MATN2), an EGF-like domain-containing protein that traps EGFR at the cell surface to facilitate its activation by EGF. We describe a pharmacological inhibitor of LOX, CCT365623, which disrupts EGFR cell surface retention and delays the growth of primary and metastatic tumour cells in vivo. Thus, we show that LOX regulates EGFR cell surface retention to drive tumour progression, and we validate the therapeutic potential of inhibiting this pathway with the small molecule inhibitor CCT365623.
Sigma-Aldrich
Anti-Lysyl Oxidase antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Anti-EGFR Antibody, neutralizing, clone LA1, clone LA1, Upstate®, from mouse
Sigma-Aldrich
Mouse IgG1 Negative Control, clone Ci4, Mouse IgG1 Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.