Merck
  • Palladium-copper nanowires-based biosensor for the ultrasensitive detection of organophosphate pesticides.
Palladium-copper nanowires-based biosensor for the ultrasensitive detection of organophosphate pesticides.
Analytica chimica acta (2017-07-25)
Dandan Song, Yan Li, Xiong Lu, Muxue Sun, Hui Liu, Guangming Yu, Faming Gao
ABSTRACT
A highly sensitive acetylcholinesterase (AChE) electrochemical biosensor for the quantitative determination of organophosphate pesticides (OPs) in vegetables and fruits based on palladium-copper nanowires (Pd-Cu NWs) was reported. AChE immobilized on the modified electrode could catalyze hydrolysis of acetylthiocholine chloride (ATCl), generating an irreversible oxidation peak. When exposed to the OPs, the activity of the AChE was inhibited and the current significantly decreased. The detection mechanism is based on the inhibition of AChE. The Pd-Cu NWs not only provide a large active surface area (0.268 ± 0.01) cm
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
Acetylthiocholine chloride, ≥99% (TLC), powder