Merck
  • AMPKα1-LDH pathway regulates muscle stem cell self-renewal by controlling metabolic homeostasis.
AMPKα1-LDH pathway regulates muscle stem cell self-renewal by controlling metabolic homeostasis.
The EMBO journal (2017-05-19)
Marine Theret, Linda Gsaier, Bethany Schaffer, Gaëtan Juban, Sabrina Ben Larbi, Michèle Weiss-Gayet, Laurent Bultot, Caterina Collodet, Marc Foretz, Dominique Desplanches, Pascual Sanz, Zizhao Zang, Lin Yang, Guillaume Vial, Benoit Viollet, Kei Sakamoto, Anne Brunet, Bénédicte Chazaud, Rémi Mounier
ABSTRACT
Control of stem cell fate to either enter terminal differentiation versus returning to quiescence (self-renewal) is crucial for tissue repair. Here, we showed that AMP-activated protein kinase (AMPK), the master metabolic regulator of the cell, controls muscle stem cell (MuSC) self-renewal. AMPKα1
Sigma-Aldrich
Calcium Phosphate Transfection Kit, Most cost effective transfection reagent kit for transient and stable transfection of DNA into mammalian cells
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Triton X-100, laboratory grade