Merck
  • Designing of macroporous magnetic bioscaffold based on functionalized methacrylate network covered by hydroxyapatites and doped with nano-MgFe
Designing of macroporous magnetic bioscaffold based on functionalized methacrylate network covered by hydroxyapatites and doped with nano-MgFe
Materials science & engineering. C, Materials for biological applications (2017-06-04)
Łukasz John, Mateusz Janeta, Sławomir Szafert
ABSTRACT
In this paper, we report on synthesis and characterization of three-dimensional biocomposite based on a polymerized 3-(trimethoxysilyl)propyl methacrylate/ethylene glycol dimethacrylate (pTMSPMA/pEGDMA) framework. The resulting composite was doped with Ca
Sigma-Aldrich
Ethyltriphenylphosphonium bromide, 99%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor