Merck
  • Megakaryocytic Smad4 Regulates Platelet Function through Syk and ROCK2 Expression.
Megakaryocytic Smad4 Regulates Platelet Function through Syk and ROCK2 Expression.
Molecular pharmacology (2017-07-01)
Yanhua Wang, Lirong Jiang, Xi Mo, Yu Lan, Xiao Yang, Xinyi Liu, Jian Zhang, Li Zhu, Junling Liu, Xiaolin Wu
ABSTRACT
Smad4, a key transcription factor in the transforming growth factor-
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
(−)-Epinephrine