Merck
Porcine B Cell Subset Responses to Toll-like Receptor Ligands.
Frontiers in immunology (2017-09-12)
Roman Othmar Braun, Sylvie Python, Artur Summerfield
ABSTRACT
Toll-like receptors (TLR) triggering of B cells are known to promote B cell expansion, differentiation of B cells into antibody-producing and memory cells, but the TLR responses of porcine B cells is poorly characterized. Therefore, this study investigated the response pattern of porcine B cell subsets to a large collection of TLR ligands and demonstrates that the TLR2 ligand Pam3Cys-SK4 and the TLR7/8 ligands gardiquimod and resiquimod are particularly efficient at inducing proliferation, CD25 and CCR7. This activation was also determined in B-cell subpopulations including a CD21
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)