Merck
  • Pet10p is a yeast perilipin that stabilizes lipid droplets and promotes their assembly.
Pet10p is a yeast perilipin that stabilizes lipid droplets and promotes their assembly.
The Journal of cell biology (2017-08-13)
Qiang Gao, Derk D Binns, Lisa N Kinch, Nick V Grishin, Natalie Ortiz, Xiao Chen, Joel M Goodman
ABSTRACT
Pet10p is a yeast lipid droplet protein of unknown function. We show that it binds specifically to and is stabilized by droplets containing triacylglycerol (TG). Droplets isolated from cells with a
Sigma-Aldrich
Cerulenin, ≥98% (HPLC), from Cephalosporium caerulens
Sigma-Aldrich
Phosphoramidon disodium salt, ≥97% (HPLC)
Sigma-Aldrich
1,2-Dioleoyl-sn-glycerol, ≥97%