Merck
  • Hypomethylation associated enhanced transcription of trefoil factor-3 mediates tamoxifen-stimulated oncogenicity of ER+ endometrial carcinoma cells.
Hypomethylation associated enhanced transcription of trefoil factor-3 mediates tamoxifen-stimulated oncogenicity of ER+ endometrial carcinoma cells.
Oncotarget (2017-11-05)
Vijay Pandey, Min Zhang, Qing-Yun Chong, Mingliang You, Ainiah Rushdiana Raquib, Amit K Pandey, Dong-Xu Liu, Liang Liu, Lan Ma, Sudhakar Jha, Zheng-Sheng Wu, Tao Zhu, Peter E Lobie
ABSTRACT
Tamoxifen (TAM) is widely used as an adjuvant therapy for women with breast cancer (BC). However, TAM possesses partial oestrogenic activity in the uterus and its use has been associated with an increased incidence of endometrial carcinoma (EC). The molecular mechanism for these observations is not well understood. Herein, we demonstrated that forced expression of
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human TFF3
Sigma-Aldrich
Stattic, ≥98% (HPLC), powder