Merck
  • Flavanols from Japanese quince (Chaenomeles japonica) fruit suppress expression of cyclooxygenase-2, metalloproteinase-9, and nuclear factor-kappaB in human colon cancer cells.
Flavanols from Japanese quince (Chaenomeles japonica) fruit suppress expression of cyclooxygenase-2, metalloproteinase-9, and nuclear factor-kappaB in human colon cancer cells.
Acta biochimica Polonica (2017-08-09)
Katarzyna Owczarek, Elżbieta Hrabec, Jakub Fichna, Dorota Sosnowska, Maria Koziołkiewicz, Jacek Szymański, Urszula Lewandowska
ABSTRACT
Natural polyphenols and polyphenol-rich extracts have been found to possess preventive and therapeutic potential against several types of cancers, including colorectal cancer (CRC), which is an example of an inflammation-associated cancer. This study examines the chemopreventive effect of a Japanese quince (Chaenomeles japonica) fruit flavanol preparation (JQFFP) on colon cancer SW-480 cells. JQFFP, rich in procyanidin monomers and oligomers, was found to inhibit the SW-480 cell viability by 40% at 150 µM catechin equivalents (CE) after 72 h incubation when compared to control, but it was non-toxic to normal colon fibroblast CCD-18Co cells. Furthermore, 100 µM CE JQFFP suppressed COX-2 mRNA expression to 36.7% of control values and protein expression to 77%. In addition, JQFFP reduced the MMP-9 protein expression (to 24% vs. control at 100 µM CE) and caused inhibition of its enzymatic activity (to 35% vs. control at 100 µM CE). Not only did JQFFP inhibit the COX-2 and MMP-9 levels, but it also reduced the NF-κB protein expression (to 65% of control) and phosphorylation of its p65 subunit (to 51%) at 100 µM CE. These results provide the first evidence that JQFFP inhibits COX-2, MMP-9, and NF-κB expression, suggesting that it has cytotoxic, anti-inflammatory, and anti-metastatic activities towards the colon cancer SW-480 cells.
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Human COX-2 ELISA Kit, for cell culture supernatants, plasma, and serum samples
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%