Merck
  • Green Tea Polyphenol EGCG Upregulates Tollip Expression by Suppressing Elf-1 Expression.
Green Tea Polyphenol EGCG Upregulates Tollip Expression by Suppressing Elf-1 Expression.
Journal of immunology (Baltimore, Md. : 1950) (2017-09-29)
Motofumi Kumazoe, Mai Yamashita, Yuki Nakamura, Kanako Takamatsu, Jaehoon Bae, Shuya Yamashita, Shuhei Yamada, Hiroaki Onda, Takashi Nojiri, Kenji Kangawa, Hirofumi Tachibana
ABSTRACT
TLR signaling is critical to innate immune system regulation; however, aberrant TLR signaling is involved in several diseases, including insulin resistance, Alzheimer's disease, and tumor metastasis. Moreover, a recent study found that TLR-4 signaling pathway inhibition might be a target for the suppression of chronic inflammatory disorders. In this article, we show that the green tea polyphenol epigallocatechin-3-O-gallate (EGCG) increases the expression of Toll interacting protein, a strong inhibitor of TLR4 signaling, by suppressing the expression of E74-like ETS transcription factor 1 (Elf-1). A mechanistic study revealed that EGCG suppressed Elf-1 expression via protein phosphatase 2A/cyclic GMP (cGMP)-dependent mechanisms. We also confirmed that orally administered EGCG and a cGMP inducer upregulated Toll interacting protein expression, increased intracellular levels of cGMP in macrophages, and suppressed Elf-1 expression. These data support EGCG and a cGMP inducer as potential candidate suppressors of TLR4 signaling.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
BAY 41-2272, ≥97% (HPLC)
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction
Sigma-Aldrich
Src Inhibitor-1, ≥98% (HPLC)