Merck
  • TIM4-TIM1 interaction modulates Th2 pattern inflammation through enhancing SIRT1 expression.
TIM4-TIM1 interaction modulates Th2 pattern inflammation through enhancing SIRT1 expression.
International journal of molecular medicine (2017-09-28)
Tianyong Hu, Xiaoqin Fan, Li Ma, Jiangqi Liu, Yunli Chang, Pingchang Yang, Shuqi Qiu, Tao Chen, Litao Yang, Zhiqiang Liu
ABSTRACT
Skewed T helper 2 (Th2)‑cell polarization plays a critical role in the pathogenesis of allergic inflammations; however, the underlying mechanisms require further elucidation. The aim of the present study was to investigate the mechanisms through which the interaction between T‑cell immunoglobulin and mucin domain (TIM)4 and TIM1 regulates the expression of silent information regulator 1 (SIRT1) in Th2 cells, and the role of SIRT1 in Th2‑cell polarization during nasal allergic inflammation. The results demonstrated that TIM4 expression by splenic dendritic cells was increased in mice with allergic rhinitis, and the TIM4̸TIM1 interaction promoted CD4+ T cells to express SIRT1 during allergic inflammation via enhancing phosphoinositide 3‑kinase/Akt phosphorylation. SIRT1 then facilitated CD4+ T‑cell proliferation through downregulating the expression of Fas ligand, caspase-3 and p53 in mice with nasal allergic inflammation. In conclusion, the interaction of TIM4̸TIM1 was found to promote Th2‑cell proliferation through enhancing SIRT1 expression in mice with nasal allergic rhinitis.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Mouse IgE ELISA Kit
Sigma-Aldrich
MISSION® esiRNA, targeting human TIMD4
Sigma-Aldrich
Sirtinol, ≥95% (HPLC)
Sigma-Aldrich
LY-294,002 hydrochloride, solid, ≥98% (HPLC)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)