Merck
In Vivo Imaging of the Programmed Death Ligand 1 by
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2017-06-08)
Dinko E González Trotter, Xiangjun Meng, Paul McQuade, Daniel Rubins, Michael Klimas, Zhizhen Zeng, Brett M Connolly, Patricia J Miller, Stacey S O'Malley, Shu-An Lin, Krista L Getty, Laurence Fayadat-Dilman, Linda Liang, Elisabet Wahlberg, Olof Widmark, Caroline Ekblad, Fredrik Y Frejd, Eric D Hostetler, Jeffrey L Evelhoch
ABSTRACT
Programmed death ligand 1 (PD-L1) is an immune regulatory ligand that binds to the T-cell immune check point programmed death 1. Tumor expression of PD-L1 is correlated with immune suppression and poor prognosis. It is also correlated with therapeutic efficacy of programmed death 1 and PD-L1 inhibitors. In vivo imaging may enable real-time follow-up of changing PD-L1 expression and heterogeneity evaluation of PD-L1 expression across tumors in the same subject. We have radiolabeled the PD-L1-binding Affibody molecule NOTA-Z