Merck
  • Evolution of Energy Related Metabolites in Plasma from Newborns with Hypoxic-Ischemic Encephalopathy during Hypothermia Treatment.
Evolution of Energy Related Metabolites in Plasma from Newborns with Hypoxic-Ischemic Encephalopathy during Hypothermia Treatment.
Scientific reports (2017-12-08)
Ángel Sánchez-Illana, Antonio Núñez-Ramiro, María Cernada, Anna Parra-Llorca, Eva Valverde, Dorotea Blanco, Maria Teresa Moral-Pumarega, Fernando Cabañas, Hector Boix, Antonio Pavon, Mercedes Chaffanel, Isabel Benavente-Fernández, Inés Tofe, Begoña Loureiro, José R Fernández-Lorenzo, Belén Fernández-Colomer, Ana García-Robles, Julia Kuligowski, Máximo Vento
ABSTRACT
Therapeutic hypothermia (TH) initiated within 6 h from birth is the most effective therapeutic approach for moderate to severe hypoxic-ischemic encephalopathy (HIE). However, underlying mechanisms and effects on the human metabolism are not yet fully understood. This work aims at studying the evolution of several energy related key metabolites in newborns with HIE undergoing TH employing gas chromatography - mass spectrometry. The method was validated following stringent FDA requirements and applied to 194 samples from a subgroup of newborns with HIE (N = 61) enrolled in a multicenter clinical trial (HYPOTOP) for the determination of lactate, pyruvate, ketone bodies and several Krebs cycle metabolites at different sampling time points. The analysis of plasma samples from newborns with HIE revealed a decrease of lactate, pyruvate and β-hydroxybutyrate concentrations, whereas rising malate concentrations were observed. In healthy control newborns (N = 19) significantly lower levels of pyruvate and lactate were found in comparison to age-matched newborns with HIE undergoing TH, whereas acetoacetate and β-hydroxybutyrate levels were clearly increased. Access to a validated analytical method and a controlled cohort of newborns with HIE undergoing hypothermia treatment for the first time allowed the in-depth study of the evolution of key metabolites of metabolic junctions in this special population.
Sigma-Aldrich
Lithium acetoacetate, ≥90% (HPLC)
Sigma-Aldrich
Sodium fumarate dibasic, ≥99%
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Methoxyamine hydrochloride, 98%