Merck
  • A CRISPR screen identifies a pathway required for paraquat-induced cell death.
A CRISPR screen identifies a pathway required for paraquat-induced cell death.
Nature chemical biology (2017-10-24)
Colleen R Reczek, Kıvanç Birsoy, Hyewon Kong, Inmaculada Martínez-Reyes, Tim Wang, Peng Gao, David M Sabatini, Navdeep S Chandel
ABSTRACT
Paraquat, a herbicide linked to Parkinson's disease, generates reactive oxygen species (ROS), which causes cell death. Because the source of paraquat-induced ROS production remains unknown, we conducted a CRISPR-based positive-selection screen to identify metabolic genes essential for paraquat-induced cell death. Our screen uncovered three genes, POR (cytochrome P450 oxidoreductase), ATP7A (copper transporter), and SLC45A4 (sucrose transporter), required for paraquat-induced cell death. Furthermore, our results revealed POR as the source of paraquat-induced ROS production. Thus, our study highlights the use of functional genomic screens for uncovering redox biology.
Sigma-Aldrich
Methyl viologen dichloride hydrate, 98%
Sigma-Aldrich
MitoPQ, ≥90% (HPLC)
Sigma-Aldrich
2,3-Dimethoxy-1,4-naphthoquinone, ≥99%, solid
Sigma-Aldrich
Methyl pyruvate, 90%, technical grade
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid