Merck
  • A reagent for covalently attaching biotin to proteins via a cleavable connector arm.
A reagent for covalently attaching biotin to proteins via a cleavable connector arm.
Archives of biochemistry and biophysics (1982-10-01)
C A Mouton, D Pang, C V Natraj, J A Shafer
Sigma-Aldrich
Sulfo-NHS-LC-LC-Biotin, ≥95%
Sigma-Aldrich
Biotin disulfide N-hydroxysuccinimide ester, >98%, powder