Merck
  • Hierarchical clustering of immunohistochemical analysis of the activated ErbB/PI3K/Akt/NF-kappaB signalling pathway and prognostic significance in prostate cancer.
Hierarchical clustering of immunohistochemical analysis of the activated ErbB/PI3K/Akt/NF-kappaB signalling pathway and prognostic significance in prostate cancer.
British journal of cancer (2010-03-11)
I H Koumakpayi, C Le Page, A-M Mes-Masson, F Saad
ABSTRACT
The PI3K/Akt signalling pathway, induced by epidermal growth factor receptor (EGFR) and Her-2, is involved in the constitutive activation of NF-kappaB in prostate cancer cell lines. In this study, we extended the in vitro observation using an ex vivo model of prostate cancer tissues and assessed the prognostic significance of the PI3K/Ak/NF-kappaB signalling determinants. We analysed a prostate cancer tissue microarray of 63 patients for the expression of total and activated EGFR, Her-2 receptors and the signalling molecules PTEN, phospho-PTEN, Akt, phospho-Akt and the NF-kappaB subunit p65. Data were analysed using Spearman's rho test, Kaplan-Meier curves and multivariate Cox regression analysis. In addition, a non-supervised hierarchical clustering analysis was applied to stratify patients according to prognostic groups in terms of risk of recurrence. The concomitant overexpression of activated EGFR and Her-2 was correlated with the nuclear expression of NF-kappaB. EGFR, phospho-EGFR, phospho-Her-2, ErbB3 and nuclear NF-kappaB were associated with the overall biochemical recurrence (BCR) of patients. The non-supervised hierarchical clustering analysis resulted in the separation of patients into five groups according to BCR. These results validate the previous in vitro data on ErbB involvement in NF-kappaB activation and shows evidence for a significant role of ErbB/PI3K/Akt/NF-kappaB signalling in the progression of prostate cancer.