Merck
LC/MS Analysis of Cetirizine and Hydroxyzine in Urine on Ascentis® Express F5
LC/MS Analysis of Cetirizine and Hydroxyzine in Urine on Ascentis® Express F5 application for LC-MS
Materials
related product
Product No.
Description
Pricing
Ascentis® Express F5, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 3 mm
Cetirizine dihydrochloride solution
1.0 mg/mL in acetonitrile: water (9:1) (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Hydroxyzine dihydrochloride solution
1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
CONDITIONS
column
Ascentis Express F5, 10 cm x 3.0 mm I.D., 2.7 μm particles (53578-U)
mobile phase
5 mM ammonium formate pH 3.0 with formic acid in 90:10, acetonitrile:water
flow rate
0.6 mL/min
pressure
1455 psi (100 bar)
column temp.
25 °C
detector
MS, ESI+, combined SIR
injection
1 μL
sample
100 ng/mL in urine dilute 1:1 with water
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany