Skip to Content
Products
Cart
0
LT
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Back
Arsenic Acid (AsV) Standard Solution
Synonym(s):
Arsenic Acid (AsV) Standard Solution, Arsenic Acid
Products
Product Category
reference materials
Manufacturer
Special Grade
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
NIST3036
Arsenic Acid (AsV) Standard Solution,
NIST
®
SRM
®
3036
Expand
View Pricing