Skip to Content
Products
Cart
0
LT
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Back
Rubidium (Rb) Standard Solution
Synonym(s):
Rubidium nitrate
Products
Product Category
reference materials
Manufacturer
Physical Form
Special Grade
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
NIST3145A
Rubidium (Rb) Standard Solution,
NIST
®
SRM
®
3145A
Expand
View Pricing