Merck
  • Synthesis of ursodeoxycholic acid from plant-source (20S)-21-hydroxy-20-methylpregn-4-en-3-one.
Synthesis of ursodeoxycholic acid from plant-source (20S)-21-hydroxy-20-methylpregn-4-en-3-one.
Steroids (2020-02-19)
Jie Wang, Xiang-Zhong Gu, Li-Ming He, Chen-Chen Li, Wen-Wei Qiu
ABSTRACT
A novel synthetic route of producing ursodeoxycholic acid (UDCA) was developed through multiple reactions from cheap and commercially available bisnoralcohol (BA). The key reaction conditions, including solvents, bases and reaction temperatures of the route were investigated and optimized. In the straightforward route for preparation of UDCA, most of the reaction steps have high conversions with average yields of 91%, and overall yield up to 59% (6 steps) from the plant-source BA. Especially in the last step of reduction and hydrolysis, there are five functional groups converted with calcd 97% per conversion in one-pot reaction. This promising route offers economical and efficient strategies for potential large-scale production of UDCA.
Sigma-Aldrich
1,3-Propylene sulfite, 99%