Merck
  • Role of growth hormone-releasing hormone in dyslipidemia associated with experimental type 1 diabetes.
Role of growth hormone-releasing hormone in dyslipidemia associated with experimental type 1 diabetes.
Proceedings of the National Academy of Sciences of the United States of America (2016-02-03)
Maritza J Romero, Rudolf Lucas, Huijuan Dou, Supriya Sridhar, Istvan Czikora, Eby M Mosieri, Ferenc G Rick, Norman L Block, Subbaramiah Sridhar, David Fulton, Neal L Weintraub, Zsolt Bagi, Andrew V Schally
ABSTRACT
Dyslipidemia associated with triglyceride-rich lipoproteins (TRLs) represents an important residual risk factor for cardiovascular and chronic kidney disease in patients with type 1 diabetes (T1D). Levels of growth hormone (GH) are elevated in T1D, which aggravates both hyperglycemia and dyslipidemia. The hypothalamic growth hormone-releasing hormone (GHRH) regulates the release of GH by the pituitary but also exerts separate actions on peripheral GHRH receptors, the functional role of which remains elusive in T1D. In a rat model of streptozotocin (STZ)-induced T1D, GHRH receptor expression was found to be up-regulated in the distal small intestine, a tissue involved in chylomicron synthesis. Treatment of T1D rats with a GHRH antagonist, MIA-602, at a dose that did not affect plasma GH levels, significantly reduced TRL, as well as markers of renal injury, and improved endothelial-dependent vasorelaxation. Glucagon-like peptide 1 (GLP-1) reduces hyperglucagonemia and postprandial TRL, the latter in part through a decreased synthesis of apolipoprotein B-48 (ApoB-48) by intestinal cells. Although plasma GLP-1 levels were elevated in diabetic animals, this was accompanied by increased rather than reduced glucagon levels, suggesting impaired GLP-1 signaling. Treatment with MIA-602 normalized GLP-1 and glucagon to control levels in T1D rats. MIA-602 also decreased secretion of ApoB-48 from rat intestinal epithelial cells in response to oleic acid stimulation in vitro, in part through a GLP-1-dependent mechanism. Our findings support the hypothesis that antagonizing the signaling of GHRH in T1D may improve GLP-1 function in the small intestine, which, in turn, diminishes TRL and reduces renal and vascular complications.
Product Description
Millipore
MILLIPLEX® Rat Metabolic Hormone Magnetic Bead Panel - Metabolism Multiplex Assay, The analytes available for this multiplex kit are: Amylin (Active), C-Peptide 2, Ghrelin (Active), GIP (Total), GLP-1 (Active), Glucagon, IL-6, Insulin, Leptin, MCP-1, PP, PYY, TNF-α.